RU
EN
Applications
Products
Services
Support
Home
Search Results
EPA 524 Rev 4 Update Mix
(1)
EPA 524 Rev 4 Update Mix
Synonyms:
EPA 524.2 Rev 4 Update Mix
EC Number:
200-659-6
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
506524
EPA 524 Rev 4 Update Mix, 200 μg/mL each component in methanol, analytical standard
Expand
View Pricing