RU
EN
Applications
Products
Services
Support
Home
Search Results
EPA 8240B/8260A GC-MS tuning mix
(2)
EPA 8240B/8260A GC-MS tuning mix
Synonyms:
4-Bromofluorobenzene
Empirical Formula (Hill Notation):
C
6
H
4
BrF
Molecular Weight:
175.00
EC Number:
200-659-6
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
47077
EPA 8240B/8260A GC-MS Tuning Mix, certified reference material, 25 μg/mL in methanol
Expand
View Pricing
47414
EPA 8240B/8260A GC-MS tuning mix, certified reference material, 25 μg/mL in methanol
Expand
View Pricing