Merck
Ethoxyacetic acid 98%

Ethoxyacetic acid

Ethoxyacetic acid 98%
Synonyms:
O-Ethylglycolic acid
Linear Formula:
C2H5OCH2COOH
CAS Number:
Molecular Weight:
104.10
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Ethoxyacetic acid, 98%