RU
EN
Applications
Products
Services
Support
Home
Search Results
Florisil
®
Adsorbent for Chromatography
(2)
Florisil
®
Adsorbent for Chromatography
Synonyms:
Florisil, Magnesium silicate, Activated magnesium silicate
Empirical Formula (Hill Notation):
MgO
3
Si
CAS Number:
1343-88-0
Molecular Weight:
100.39
EC Number:
215-681-1
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
46385
Florisil
®
Adsorbent for Chromatography, 60-100 mesh
Expand
View Pricing
46384
Florisil
®
Adsorbent for Chromatography, 30-60 mesh
Expand
View Pricing