Hydrochloric acid 32% EMPLURA®

Hydrochloric acid 32%

Hydrochloric acid 32% EMPLURA®
Synonyms:
Hydrogen chloride solution, Hydrochloric acid solution
Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Hydrochloric acid 32%, for analysis EMSURE®
Hydrochloric acid 32%, EMPLURA®