Products
RU
EN
Products
Applications
Products
Services
Support
Home
Search Results
Lansoprazole Related Compound B
(2)
Lansoprazole Related Compound B
Synonyms:
2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-pyridin-2-yl]methyl]sulfanyl]-1
H
-benzimidazole, Lansoprazole sulfide
Empirical Formula (Hill Notation):
C
16
H
14
F
3
N
3
OS
CAS Number:
103577-40-8
Molecular Weight:
353.36
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1356931
Lansoprazole Related Compound B, United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR2956
Lansoprazole Related Compound B, certified reference material, pharmaceutical secondary standard
Expand
View Pricing