RU
EN
Applications
Products
Services
Support
Home
Search Results
MG-132, ≥95% by HPLC
(1)
MG-132, ≥95% by HPLC
Synonyms:
Z-Leu-Leu-Leu-al, MG-132
Empirical Formula (Hill Notation):
C
26
H
41
N
3
O
5
CAS Number:
133407-82-6
Molecular Weight:
475.62
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
474787
MG-132, ≥95% by HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (K
i
= 4 nM).
Expand
View Pricing