N-(1-Naphthyl)ethylenediamine dihydrochloride GR for analysis ACS

N-(1-Naphthyl)ethylenediamine dihydrochloride

N-(1-Naphthyl)ethylenediamine dihydrochloride GR for analysis ACS
Synonyms:
2-(1-Naphthylamino)ethylamine dihydrochloride, N-(1-Naphthyl)ethylenediamine dihydrochloride
Linear Formula:
C10H7NHCH2CH2NH2 · 2HCl
Molecular Weight:
259.17

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, >98%
N-(1-Naphthyl)ethylenediamine dihydrochloride, ≥98%
N-(1-Naphthyl)ethylenediamine dihydrochloride, for determination of sulfonamide and nitrite, ACS reagent, ≥98%
N-(1-Naphthyl)ethylenediamine dihydrochloride, GR for analysis ACS

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.