Nicotinic acid extra pure Ph Eur,BP,USP

Nicotinic acid

Nicotinic acid extra pure Ph Eur,BP,USP
Synonyms:
Pellagra preventive factor, Niacin, Pyridine-3-carboxylic acid, Vitamin B3, 3-Picolinic acid, Nicotinic acid
Linear Formula:
3-(COOH)-C5H4N
CAS Number:
Molecular Weight:
123.11

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Nicotinic acid, ≥98%
Nicotinic acid, ≥99.5% (HPLC)
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Nicotinic acid, analytical standard
Nicotinic acid, meets USP testing specifications
Nicotinic acid, certified reference material, TraceCERT®
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard
Nicotinic acid, for inorganic trace analysis, ≥99.999% (metals basis)
Nicotinic acid, for synthesis
Nicotinic acid, extra pure Ph Eur,BP,USP

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.