Merck
Potassium chloride solution (nominal 12.8 mS/cm) reference material for the measurement of electrolytic conductivity, traceable to PTB (c=0.1 mol/l) Certipur®

Potassium chloride solution (nominal 12.8 mS/cm)

Potassium chloride solution (nominal 12.8 mS/cm) reference material for the measurement of electrolytic conductivity, traceable to PTB (c=0.1 mol/l) Certipur®
Synonyms:
Potassium chloride solution
Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Potassium chloride solution (nominal 12.8 mS/cm), certified reference material for the measurement of electrolytic conductivity (c=0.1 mol/l) Certipur®
Potassium chloride solution (nominal 12.8 mS/cm), reference material for the measurement of electrolytic conductivity, traceable to PTB (c=0.1 mol/l) Certipur®