Merck
  • Inflammation-activated CXCL16 pathway contributes to tubulointerstitial injury in mouse diabetic nephropathy.
Inflammation-activated CXCL16 pathway contributes to tubulointerstitial injury in mouse diabetic nephropathy.
Acta pharmacologica Sinica (2018-04-06)
Ze-Bo Hu, Kun-Ling Ma, Yang Zhang, Gui-Hua Wang, Liang Liu, Jian Lu, Pei-Pei Chen, Chen-Chen Lu, Bi-Cheng Liu
ABSTRACT
Inflammation and lipid disorders play crucial roles in synergistically accelerating the progression of diabetic nephropathy (DN). In this study we investigated how inflammation and lipid disorders caused tubulointerstitial injury in DN in vivo and in vitro. Diabetic db/db mice were injected with 10% casein (0.5 mL, sc) every other day for 8 weeks to cause chronic inflammation. Compared with db/db mice, casein-injected db/db mice showed exacerbated tubulointerstitial injury, evidenced by increased secretion of extracellular matrix (ECM) and cholesterol accumulation in tubulointerstitium, which was accompanied by activation of the CXC chemokine ligand 16 (CXCL16) pathway. In the in vitro study, we treated HK-2 cells with IL-1β (5 ng/mL) and high glucose (30 mmol/L). IL-1β treatment increased cholesterol accumulation in HK-2 cells, leading to greatly increased ROS production, ECM protein expression levels, which was accompanied by the upregulated expression levels of proteins in the CXCL16 pathway. In contrast, after CXCL16 in HK-2 cells was knocked down by siRNA, the IL-1β-deteriorated changes were attenuated. In conclusion, inflammation accelerates renal tubulointerstitial lesions in mouse DN via increasing the activity of CXCL16 pathway.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human CXCL16
Pricing and availability is not currently available.