  • Search Results
  • Microcasting with agarose gel via degassed polydimethylsiloxane molds for repellency-guided cell patterning
Microcasting with agarose gel via degassed polydimethylsiloxane molds for repellency-guided cell patterning
Royal Society of Chemistry Advances (2016)
Tanaka N, et al.