RU
EN
Applications
Products
Services
Support
Home
Search Results
Use of within-group designs to test anti-tussive drugs in conscious guinea-pigs
Use of within-group designs to test anti-tussive drugs in conscious guinea-pigs
Journal of Pharmacological and Toxicological Methods (2010)
Venkatasamy R, et al.