  • Search Results
  • Generation of an iPSC line from a retinitis pigmentosa patient carrying a homozygous mutation in CERKL and a healthy sibling.
Generation of an iPSC line from a retinitis pigmentosa patient carrying a homozygous mutation in CERKL and a healthy sibling.
Stem cell research (2019-05-15)
Arantxa Bolinches-Amorós, Marian León, Verónica Del Buey Furió, Gemma Marfany, Roser Gonzàlez-Duarte, Slaven Erceg, Dunja Lukovic
ABSTRACT
Dermal fibroblasts from an autosomal recessive retinitis pigmentosa (RP) patient, homozygous for the mutation c.769 C>T, p.Arg257Ter, in CERKL (Ceramide Kinase-Like) gene, and a healthy sibling were derived and reprogrammed by Sendai virus. The generated human induced pluripotent stem cell (hiPSC) lines RP3-FiPS4F1 and Ctrl3-FiPS4F1, were free of genomically integrated reprogramming genes, showed stable karyotypes, expressed pluripotency markers and could be differentiated towards the three germ layers in vitro. These hiPSC lines offer a useful resource to study RP pathomechanisms, drug testing and therapeutic opportunities.