  • Search Results
  • The Tumor Suppressor FBW7 and the Vitamin D Receptor Are Mutual Cofactors in Protein Turnover and Transcriptional Regulation.
The Tumor Suppressor FBW7 and the Vitamin D Receptor Are Mutual Cofactors in Protein Turnover and Transcriptional Regulation.
Molecular cancer research : MCR (2019-01-05)
Reyhaneh Salehi-Tabar, Babak Memari, Hilary Wong, Vassil Dimitrov, Natacha Rochel, John H White
ABSTRACT
The E3 ligase and tumor suppressor FBW7 targets drivers of cell-cycle progression such as the oncogenic transcription factor c-MYC, for proteasomal degradation. Vitamin D signaling regulates c-MYC expression and turnover in vitro and in vivo, which is highly significant as epidemiologic data link vitamin D deficiency to increased cancer incidence. We hypothesized that FBW7 and the vitamin D receptor (VDR) controlled each other's function as regulators of protein turnover and gene transcription, respectively. We found that hormonal 1,25-dihydroxyvitamin D3 (1,25D) rapidly enhanced the interaction of FBW7 with VDR and with c-MYC, whereas it blocked FBW7 binding to c-MYC antagonist MXD1. 1,25D stimulated the recruitment of FBW7, SCF complex subunits, and ubiquitin to DNA-bound c-MYC, consistent with 1,25D-regulated c-MYC degradation on DNA. 1,25D also accelerated the turnover of other FBW7 target proteins such as Cyclin E, c-JUN, MCL1, and AIB1, and, importantly, FBW7 depletion attenuated the 1,25D-induced cell-cycle arrest. Although the VDR contains a consensus FBW7 recognition motif in a VDR-specific insertion domain, its mutation did not affect FBW7-VDR interactions, and FBW7 ablation did not stabilize the VDR. Remarkably, however, FBW7 is essential for optimal VDR gene expression. In addition, the FBW7 and SCF complex subunits are recruited to 1,25D-induced genes and FBW7 depletion inhibited the 1,25D-dependent transactivation. Collectively, these data show that the VDR and FBW7 are mutual cofactors, and provide a mechanistic basis for the cancer-preventive actions of vitamin D. IMPLICATIONS: The key findings show that the VDR and the E3 ligase FBW7 regulate each other's functions in transcriptional regulation and control of protein turnover, respectively, and provide a molecular basis for cancer-preventive actions of vitamin D.Visual Overview: http://mcr.aacrjournals.org/content/17/3/709/F1.large.jpg.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human FBXW7
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.