  • Search Results
  • Transcription Restart Establishes Chromatin Accessibility after DNA Replication.
Transcription Restart Establishes Chromatin Accessibility after DNA Replication.
Molecular cell (2019-05-28)
Kathleen R Stewart-Morgan, Nazaret Reverón-Gómez, Anja Groth
ABSTRACT
DNA replication is highly disruptive to chromatin, leading to eviction of nucleosomes, RNA polymerase, and regulatory factors. When and how transcription resumes on DNA following DNA replication is unknown. Here we develop a replication-coupled assay for transposase-accessible chromatin (repli-ATAC-seq) to investigate active chromatin restoration post-replication in mouse embryonic stem cells. We find that nascent chromatin is inaccessible and transcriptionally silenced, with accessibility and RNA polymerase occupancy re-appearing within 30 minutes. Chromatin accessibility restores differentially genome wide, with super enhancers regaining transcription factor occupancy faster than other genomic features. We also identify opportunistic and transiently accessible chromatin within gene bodies after replication. Systematic inhibition of transcription shows that transcription restart is required to re-establish active chromatin states genome wide and resolve opportunistic binding events resulting from DNA replication. Collectively, this establishes a central role for transcription in overcoming the genome-wide chromatin inaccessibility imposed by DNA replication every cell division.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tris(3-hydroxypropyltriazolylmethyl)amine, 95%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
Triptolide, from Tripterygium wilfordii, ≥98% (HPLC), solid
Sigma-Aldrich
Shields and Sang M3 Insect Medium, With L-glutamine, without potassium bicarbonate, powder, suitable for insect cell culture
Sigma-Aldrich
Select Yeast Extract, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Potassium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, E501, 99.5-101.0% (acidimetric)