  • Search Results
  • Gastrodin attenuates proliferation and inflammatory responses in activated microglia through Wnt/β-catenin signaling pathway.
Gastrodin attenuates proliferation and inflammatory responses in activated microglia through Wnt/β-catenin signaling pathway.
Brain research (2019-04-27)
Yue-Yi Yao, Li-Gong Bian, Ping Yang, Yue Sui, Run Li, Yuan-Li Chen, Lin Sun, Qing-Long Ai, Lian-Mei Zhong, Di Lu
ABSTRACT
Microglia contribute to the regulation of neuroinflammation and play an important role in the pathogenesis of brain disorders. Thus, regulation of neuroinflammation triggered by activation of microglia has become a promising therapeutic strategy. Here, we investigated the beneficial effects of Gastrodin in activated microglia and analyzed the underlying molecular mechanisms. Microglia activation was regulated by Gastrodin not only in terms of microglia population size but also production of inflammatory mediators. Gastrodin inhibited the expression of inducible nitric oxide synthase (iNOS), tumor necrosis factor-α (TNF-α), cyclin-D1 and Ki67 in lipopolysaccharide (LPS)-stimulated BV-2 or primary microglia. Gastrodin also suppressed the expression of iNOS and Ki67 in activated microglia in three-day-old LPS-injected postnatal rats. In addition, the present results have shown that Gastrodin inhibited LPS-induced phosphorylation of glycogen synthase kinase-3β (GSK-3β) at Ser 9 and β-catenin activity. We further extended our investigation to determine whether Wnt/β-catenin signaling pathway was involved in the anti-inflammatory and anti-proliferation function of Gastrodin. β-Catenin antagonist (XAV939) was used to block LPS-mediated upregulation of iNOS, TNF-α, cyclin-D1, nitric oxide (NO) and the number of cells in the G2/M+S phase of cell cycle. Moreover, treatment with LiCl, a special Wnt/β-catenin pathway agonist significantly blocked Gastrodin-mediated down-regulation of iNOS, TNF-α, cyclin-D1, NO and the number of cells in the G2/M+S phase of cell cycle in LPS-stimulated BV-2 microglia. Taken together, the present results suggested that Gastrodin mediated anti-inflammatory and anti-proliferation effects in activated microglia by modulating the Wnt/β-catenin signaling pathway.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Fluoroshield with DAPI, histology mounting medium
Sigma-Aldrich
Lectin from Lycopersicon esculentum (tomato), FITC conjugate, buffered aqueous solution
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–Cy3 antibody produced in sheep, affinity isolated antibody, buffered aqueous solution