  • Search Results
  • Cross-Regulation between TDP-43 and Paraspeckles Promotes Pluripotency-Differentiation Transition.
Cross-Regulation between TDP-43 and Paraspeckles Promotes Pluripotency-Differentiation Transition.
Molecular cell (2019-05-03)
Miha Modic, Markus Grosch, Gregor Rot, Silvia Schirge, Tjasa Lepko, Tomohiro Yamazaki, Flora C Y Lee, Ejona Rusha, Dmitry Shaposhnikov, Michael Palo, Juliane Merl-Pham, Davide Cacchiarelli, Boris Rogelj, Stefanie M Hauck, Christian von Mering, Alexander Meissner, Heiko Lickert, Tetsuro Hirose, Jernej Ule, Micha Drukker
ABSTRACT
RNA-binding proteins (RBPs) and long non-coding RNAs (lncRNAs) are key regulators of gene expression, but their joint functions in coordinating cell fate decisions are poorly understood. Here we show that the expression and activity of the RBP TDP-43 and the long isoform of the lncRNA Neat1, the scaffold of the nuclear compartment "paraspeckles," are reciprocal in pluripotent and differentiated cells because of their cross-regulation. In pluripotent cells, TDP-43 represses the formation of paraspeckles by enhancing the polyadenylated short isoform of Neat1. TDP-43 also promotes pluripotency by regulating alternative polyadenylation of transcripts encoding pluripotency factors, including Sox2, which partially protects its 3' UTR from miR-21-mediated degradation. Conversely, paraspeckles sequester TDP-43 and other RBPs from mRNAs and promote exit from pluripotency and embryonic patterning in the mouse. We demonstrate that cross-regulation between TDP-43 and Neat1 is essential for their efficient regulation of a broad network of genes and, therefore, of pluripotency and differentiation.
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Poly(ethylene glycol), average Mn 400
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Roche
DIG RNA Labeling Kit (SP6/T7)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Alkaline Phosphatase Blue Membrane Substrate Solution, sufficient for 400 mL working substrate
Sigma-Aldrich
Anti-TDP-43 (N-terminal region) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human TARDBP
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.