  • Search Results
  • Interplay between interferon regulatory factor 1 and BRD4 in the regulation of PD-L1 in pancreatic stellate cells.
Interplay between interferon regulatory factor 1 and BRD4 in the regulation of PD-L1 in pancreatic stellate cells.
Scientific reports (2018-09-07)
Kazumi Ebine, Krishan Kumar, Thao N Pham, Mario A Shields, Katharine A Collier, Meng Shang, Brian T DeCant, Raul Urrutia, Rosa F Hwang, Sam Grimaldo, Daniel R Principe, Paul J Grippo, David J Bentrem, Hidayatullah G Munshi
ABSTRACT
The fibrotic reaction is a characteristic feature of human pancreatic ductal adenocarcinoma (PDAC) tumors. It is associated with activation and proliferation of pancreatic stellate cells (PSCs), which are key regulators of fibrosis in vivo. While there is increasing interest in the regulation of PD-L1 expression in cancer and immune cells, the expression and regulation of PD-L1 in other stromal cells, such as PSCs, has not been fully evaluated. Here we show that PSCs in vitro express higher PD-L1 mRNA and protein levels compared to the levels present in PDAC cells. We show that inhibitors targeting bromodomain and extra-terminal (BET) proteins and BRD4 knockdown decrease interferon-γ (IFN-γ)-induced PD-L1 expression in PSCs. We also show that c-MYC, one of the well-established targets of BET inhibitors, does not mediate IFN-γ-regulated PD-L1 expression in PSCs. Instead we show that interferon regulatory factor 1 (IRF1) mediates IFN-γ-induced PD-L1 expression in PSCs. Finally, while we show that BET inhibitors do not regulate IFN-γ-induced IRF1 expression in PSCs, BET inhibitors decrease binding of IRF1 and BRD4 to the PD-L1 promoter. Together, these results demonstrate the interplay between IRF1 and BRD4 in the regulation of PD-L1 in PSCs.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
KiCqStart® SYBR® Green qPCR ReadyMix, with ROX for ABI instruments
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution