  • Search Results
  • Anti-glycation, anti-hemolysis, and ORAC activities of demethylcurcumin and tetrahydroxycurcumin in vitro and reductions of oxidative stress in D-galactose-induced BALB/c mice in vivo.
Anti-glycation, anti-hemolysis, and ORAC activities of demethylcurcumin and tetrahydroxycurcumin in vitro and reductions of oxidative stress in D-galactose-induced BALB/c mice in vivo.
Botanical studies (2019-06-30)
Yuh-Hwa Liu, Tai-Lin Lee, Chuan-Hsiao Han, Yi-Shan Lee, Wen-Chi Hou
ABSTRACT
There were few report concerning anti-glycation and antioxidant activities of the minor amounts of components in curcuminoids, demethylcurcumin and tetrahydroxycurcumin, in vitro and in vivo. The bovine serum albumin/galactose of non-enzymatic glycation models, radical-induced hemolysis, and oxygen radical absorbance capacity (ORAC) were studied in vitro, and the D-galactose-induced oxidative stress in BALB/c mice and then demethylcurcumin or tetrahydroxycurcumin interventions in vivo. The parameters of oxidative stress in plasma and brain extracts were determined among animal groups with or without both curcuminoids interventions. The demethylcurcumin and tetrahydroxycurcumin exhibited anti-glycation, anti-hemolysis, and ORAC activities, and showed much better and significant difference (P < 0.05) compared to those of curcumin in vitro. In animal experiments, the intervened two curcuminoids at both concentrations showed to lower serum malondialdehyde (MDA), brain MDA levels and iNOS protein expressions, and elevate serum ORAC activities, and showed difference (P < 0.05) compared to the galactose-induced control. The demethylcurcumin and tetrahydroxycurcumin showed potentials in developing functional foods for antioxidant-related purposes.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution