  • Search Results
  • Synthesis of molecularly imprinted polymers using a functionalized initiator for chiral-selective recognition of propranolol.
Synthesis of molecularly imprinted polymers using a functionalized initiator for chiral-selective recognition of propranolol.
Chirality (2020-01-14)
Weifeng Liu, Clovia Holdsworth, Lei Ye
ABSTRACT
We present a new concept of synthesis for preparation of molecularly imprinted polymers using a functionalized initiator to replace the traditional functional monomer. Using propranolol as a model template, a carboxyl-functionalized radical initiator was demonstrated to lead to high-selectivity polymer particles prepared in a standard precipitation polymerization system. When a single enantiomer of propranolol was used as template, the imprinted polymer particles exhibited clear chiral selectivity in an equilibrium binding experiment. Unlike the previous molecular imprinting systems where the active free radicals can be distant from the template-functional monomer complex, the method reported in this work makes sure that the actual radical polymerization takes place in the vicinity of the template-associated functional groups. The success of using functional initiator to synthesize molecularly imprinted polymers brings in new possibilities to improve the functional performance of molecularly imprinted synthetic receptors.
Sigma-Aldrich
Trimethylolpropane trimethacrylate, contains 250 ppm monomethyl ether hydroquinone as inhibitor, technical grade
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder