  • Search Results
  • Baicalin relieves inflammation stimulated by lipopolysaccharide via upregulating TUG1 in liver cells.
Baicalin relieves inflammation stimulated by lipopolysaccharide via upregulating TUG1 in liver cells.
Journal of physiology and biochemistry (2019-08-10)
Yanqiu Huang, Mengyan Sun, Xuefang Yang, Aiyu Ma, Yujie Ma, Aiying Zhao
ABSTRACT
Hepatitis has become a major social, health, and economic problem worldwide. Herein, we tested the beneficial influence of baicalin, a flavonoid extracted from the roots of Scutellaria baicalensis, on human normal liver L-02 and THLE2 cell apoptosis and inflammatory reaction stimulated by lipopolysaccharide (LPS) and possible molecular mechanisms. L-02 and THLE2 cell viability and apoptosis after LPS and/or baicalin treatment were tested using CCK-8 assay and Annexin V-FITC/PI apoptosis kit, respectively. qRT-PCR was used to measure the MCP-1, IL-6, TNF-α, and lncRNA taurine upregulated gene 1 (TUG1) expressions in L-02 and THLE2 cells. sh-TUG1 was transfected to knockdown TUG1. SB203580 was used as inhibitor of p38MAPK pathway, while SP600125 was used as inhibitor of JNK pathway. We discovered that LPS stimulation caused L-02 and THLE2 cell apoptosis and inflammatory reaction. Baicalin relieved the L-02 and THLE2 cell apoptosis and inflammatory reaction stimulated by LPS. Moreover, LPS lowered the TUG1 expression in L-02 cells, while baicalin promoted the TUG1 expression in L-02 and L-02 and THLE2 cells, as well as inactivated p38MAPK and JNK pathways in LPS-stimulated L-02 cells. Besides, knockdown of TUG1 activated p38MAPK and JNK pathways and promoted inflammatory cytokine expression in L-02 cells. In conclusion, this study further affirmed the beneficial influences of baicalin on LPS-stimulated human normal liver cell apoptosis and inflammatory reaction. Baicalin relived liver cell inflammation stimulated by LPS might be via upregulating TUG1 and then inactivating p38MAPK and JNK pathways.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
StableCell DMEM - high glucose, With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Baicalin, 95%