  • Search Results
  • Gliomas Interact with Non-glioma Brain Cells via Extracellular Vesicles.
Gliomas Interact with Non-glioma Brain Cells via Extracellular Vesicles.
Cell reports (2020-02-27)
Xiaofei Gao, Zhaohuan Zhang, Tomoyuki Mashimo, Bo Shen, James Nyagilo, Hao Wang, Yihui Wang, Zhida Liu, Aditi Mulgaonkar, Xiao-Ling Hu, Sara G M Piccirillo, Ugur Eskiocak, Digant P Davé, Song Qin, Yongjie Yang, Xiankai Sun, Yang-Xin Fu, Hui Zong, Wenzhi Sun, Robert M Bachoo, Woo-Ping Ge
ABSTRACT
Emerging evidence suggests that crosstalk between glioma cells and the brain microenvironment may influence brain tumor growth. To date, known reciprocal interactions among these cells have been limited to the release of paracrine factors. Combining a genetic strategy with longitudinal live imaging, we find that individual gliomas communicate with distinct sets of non-glioma cells, including glial cells, neurons, and vascular cells. Transfer of genetic material is achieved mainly through extracellular vesicles (EVs), although cell fusion also plays a minor role. We further demonstrate that EV-mediated communication leads to the increase of synaptic activity in neurons. Blocking EV release causes a reduction of glioma growth in vivo. Our findings indicate that EV-mediated interaction between glioma cells and non-glioma brain cells alters the tumor microenvironment and contributes to glioma development.
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Triton X-100, laboratory grade