  • Search Results
  • KAT3-dependent acetylation of cell type-specific genes maintains neuronal identity in the adult mouse brain.
KAT3-dependent acetylation of cell type-specific genes maintains neuronal identity in the adult mouse brain.
Nature communications (2020-05-24)
Michal Lipinski, Rafael Muñoz-Viana, Beatriz Del Blanco, Angel Marquez-Galera, Juan Medrano-Relinque, José M Caramés, Andrzej A Szczepankiewicz, Jordi Fernandez-Albert, Carmen M Navarrón, Roman Olivares, Grzegorz M Wilczyński, Santiago Canals, Jose P Lopez-Atalaya, Angel Barco
ABSTRACT
The lysine acetyltransferases type 3 (KAT3) family members CBP and p300 are important transcriptional co-activators, but their specific functions in adult post-mitotic neurons remain unclear. Here, we show that the combined elimination of both proteins in forebrain excitatory neurons of adult mice resulted in a rapidly progressing neurological phenotype associated with severe ataxia, dendritic retraction and reduced electrical activity. At the molecular level, we observed the downregulation of neuronal genes, as well as decreased H3K27 acetylation and pro-neural transcription factor binding at the promoters and enhancers of canonical neuronal genes. The combined deletion of CBP and p300 in hippocampal neurons resulted in the rapid loss of neuronal molecular identity without de- or transdifferentiation. Restoring CBP expression or lysine acetylation rescued neuronal-specific transcription in cultured neurons. Together, these experiments show that KAT3 proteins maintain the excitatory neuron identity through the regulation of histone acetylation at cell type-specific promoter and enhancer regions.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-HIST2H2AC antibody produced in rabbit, IgG fraction of antiserum