  • Search Results
  • Facilitated recruitment of Pdc2p, a yeast transcriptional activator, in response to thiamin starvation.
Facilitated recruitment of Pdc2p, a yeast transcriptional activator, in response to thiamin starvation.
FEMS microbiology letters (2012-03-13)
Kazuto Nosaka, Hiroyoshi Esaki, Mari Onozuka, Hiroyuki Konno, Yasunao Hattori, Kenichi Akaji
ABSTRACT
In Saccharomyces cerevisiae, genes involved in thiamin pyrophosphate (TPP) synthesis (THI genes) and the pyruvate decarboxylase structural gene PDC5 are transcriptionally induced in response to thiamin starvation. Three positive regulatory factors (Thi2p, Thi3p, and Pdc2p) are involved in the expression of THI genes, whereas only Pdc2p is required for the expression of PDC5. Thi2p and Pdc2p serve as transcriptional activators and each factor can interact with Thi3p. The target consensus DNA sequence of Thi2p has been deduced. When TPP is not bound to Thi3p, the interactions between the regulatory factors are increased and THI gene expression is upregulated. In this study, we demonstrated that Pdc2p interacts with the upstream region of THI genes and PDC5. The association of Pdc2p or Thi2p with THI gene promoters was enhanced by thiamin starvation, suggesting that Pdc2p and Thi2p assist each other in their recruitment to the THI promoters via interaction with Thi3p. It is highly likely that, under thiamin-deprived conditions, a ternary Thi2p/Thi3p/Pdc2p complex is formed and transactivates THI genes in yeast cells. On the other hand, the association of Pdc2p with PDC5 was unaffected by thiamin. We also identified a DNA element in the upstream region of PDC5, which can bind to Pdc2p and is required for the expression of PDC5.
Sigma-Aldrich
Thiamine pyrophosphate, ≥95%