  • Search Results
  • Coinfection by Trypanosoma cruzi and a fungal pathogen increases survival of Chagasic bugs: advice against a fungal control strategy.
Coinfection by Trypanosoma cruzi and a fungal pathogen increases survival of Chagasic bugs: advice against a fungal control strategy.
Bulletin of entomological research (2019-11-07)
A Laura Flores-Villegas, Margarita Cabrera-Bravo, José A De Fuentes-Vicente, J Guillermo Jiménez-Cortés, Paz María Salazar-Schettino, Martha Irene Bucio-Torres, Alex Córdoba-Aguilar
ABSTRACT
Triatomine bugs carry the parasitic protozoa Trypanosoma cruzi, the causal agent of Chagas disease. It is known that both the parasite and entomopathogenic fungi can decrease bug survival, but the combined effect of both pathogens is not known, which is relevant for biological control purposes. Herein, the survival of the triatomine Meccus pallidipennis (Stal, 1872) was compared when it was coinfected with the fungus Metarhizium anisopliae (Metschnikoff) and T. cruzi, and when both pathogens acted separately. The immune response of the insect was also studied, using phenoloxidase activity in the bug gut and hemolymph, to understand our survival results. Contrary to expectations, triatomine survival was higher in multiple than in single challenges, even though the immune response was lower in cases of multiple infection. We postulate that T. cruzi exerts a protective effect and/or that the insect reduced the resources allocated to defend itself against both pathogens. Based on the present results, the use of M. anisopliae as a control agent should be re-considered.
Product Description
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile