  • Search Results
  • Molecular Stressors Engender Protein Connectivity Dysfunction through Aberrant N-Glycosylation of a Chaperone.
Molecular Stressors Engender Protein Connectivity Dysfunction through Aberrant N-Glycosylation of a Chaperone.
Cell reports (2020-07-02)
Pengrong Yan, Hardik J Patel, Sahil Sharma, Adriana Corben, Tai Wang, Palak Panchal, Chenghua Yang, Weilin Sun, Thais L Araujo, Anna Rodina, Suhasini Joshi, Kenneth Robzyk, Srinivasa Gandu, Julie R White, Elisa de Stanchina, Shanu Modi, Yelena Y Janjigian, Elizabeth G Hill, Bei Liu, Hediye Erdjument-Bromage, Thomas A Neubert, Nanette L S Que, Zihai Li, Daniel T Gewirth, Tony Taldone, Gabriela Chiosis
ABSTRACT
Stresses associated with disease may pathologically remodel the proteome by both increasing interaction strength and altering interaction partners, resulting in proteome-wide connectivity dysfunctions. Chaperones play an important role in these alterations, but how these changes are executed remains largely unknown. Our study unveils a specific N-glycosylation pattern used by a chaperone, Glucose-regulated protein 94 (GRP94), to alter its conformational fitness and stabilize a state most permissive for stable interactions with proteins at the plasma membrane. This "protein assembly mutation' remodels protein networks and properties of the cell. We show in cells, human specimens, and mouse xenografts that proteome connectivity is restorable by inhibition of the N-glycosylated GRP94 variant. In summary, we provide biochemical evidence for stressor-induced chaperone-mediated protein mis-assemblies and demonstrate how these alterations are actionable in disease.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Benzenesulfonic acid, technical, ≥90% (T)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit
Sigma-Aldrich
Anti-GRP94 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), peroxidase conjugate, lyophilized powder
Sigma-Aldrich
Anti-phospho-RELA (pSer529) antibody produced in rabbit, affinity isolated antibody