  • Search Results
  • Discovery of 2-aryl and 2-pyridinylbenzothiazoles endowed with antimicrobial and aryl hydrocarbon receptor agonistic activities.
Discovery of 2-aryl and 2-pyridinylbenzothiazoles endowed with antimicrobial and aryl hydrocarbon receptor agonistic activities.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2020-05-30)
Elizabeth Goya-Jorge, Fatma Abdmouleh, Laureano E Carpio, Rosa M Giner, Maité Sylla-Iyarreta Veitía
ABSTRACT
Benzothiazole is a privileged scaffold in medicinal chemistry present in diverse bioactive compounds with multiple pharmacological applications such as analgesic, anticonvulsant, antidiabetic, anti-inflammatory, anticancer and radioactive amyloidal imagining agents. We reported in this work the study of sixteen functionalized 2-aryl and 2-pyridinylbenzothiazoles as antimicrobial agents and as aryl hydrocarbon receptor (AhR) modulators. The antimicrobial activity against Gram-positive (S. aureus and M. luteus) and Gram-negative (P. aeruginosa, S. enterica and E. coli) pathogens yielded MIC ranging from 3.13 to 50 μg/mL and against the yeast C. albicans, the benzothiazoles displayed MIC from 12.5 to 100 μg/mL. All compounds showed promising antibiofilm activity against S. aureus and P. aeruginosa. The arylbenzothiazole 12 displayed the greatest biofilm eradication in S. aureus (74%) subsequently verified by fluorescence microscopy. The ability of benzothiazoles to modulate AhR expression was evaluated in a cell-based reporter gene assay. Six benzothiazoles (7, 8-10, 12, 13) induced a significant AhR-mediated transcription and interestingly compound 12 was also the strongest AhR-agonist identified. Structure-activity relationships are suggested herein for the AhR-agonism and antibiofilm activities. Furthermore, in silico predictions revealed a good ADMET profile and druglikeness for the arylbenzothiazole 12 as well as binding similarities to AhR compared with the endogenous agonist FICZ.
Product Description
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile