  • Search Results
  • Molecular Motions and Interactions in Aqueous Solutions of Thymosin-β4 , Stabilin CTD and Their 1 : 1 Complex, Studied by 1 H-NMR Spectroscopy.
Molecular Motions and Interactions in Aqueous Solutions of Thymosin-β4 , Stabilin CTD and Their 1 : 1 Complex, Studied by 1 H-NMR Spectroscopy.
Chemphyschem : a European journal of chemical physics and physical chemistry (2020-05-30)
M Bokor, Á Tantos, A Mészáros, B Jenei, R Haminda, P Tompa, K Tompa
ABSTRACT
Wide-line 1 H NMR measurements were extended and all results were interpreted in a thermodynamics-based new approach on aqueous solutions of thymosin-β4 (Tβ4 ), stabilin cytoplasmic domain (CTD), and their 1 : 1 complex. Energy distributions of potential barriers controlling the motion of protein-bound water molecules were determined. Heterogeneous and homogeneous regions were found in the protein-water interface. The measure of heterogeneity of this interface gives quantitative value for the portion of disordered parts in the protein. Ordered structural elements were found extending up to ∼20 % of the individual whole proteins. About 40 % of the binding sites of free Tβ4 get involved in bonds holding the complex together. The complex has the most heterogeneous solvent accessible surface (SAS) in terms of protein-water interactions. The complex is more disordered than Tβ4 or stabilin CTD. The greater SAS area of the complex is interpreted as a clear sign of its open structure.
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%