  • Search Results
  • Characterization of endogenous fluorescence in nonsmall lung cancerous cells: A comparison with nonmalignant lung normal cells.
Characterization of endogenous fluorescence in nonsmall lung cancerous cells: A comparison with nonmalignant lung normal cells.
Journal of biophotonics (2020-02-19)
Kamlesh Awasthi, Feng-Lin Chang, Pei-Ying Hsieh, Hsin-Yun Hsu, Nobuhiro Ohta
ABSTRACT
Monitoring fluorescence properties of endogenous fluorophores such as nicotinamide adenine dinucleotide (NADH) and flavin adenine dinucleotide (FAD) in normal and cancerous cells provide substantial information noninvasively on biochemical and biophysical aspects of metabolic dysfunction of cancerous cells. Time-resolved spectral profiles and fluorescence lifetime images of NADH and FAD were obtained in human lung nonsmall carcinomas (H661 and A549) and normal lung cells (MRC-5). Both fluorophores show the fast and slowly decaying emission components upon pulsed excitation, and fluorescence spectra of NADH and FAD show blue- and red-shifts, respectively, during their decay. All identified lifetime components of NADH and FAD were found to be shorter in cancerous cells than in normal cells, no matter how they were measured under different extra-cellular conditions (cells suspended in cuvette and cells attached on glass substrate), indicating that the changes in metabolism likely altered the subcellular milieu and potentially also affected the interaction of NADH and FAD with enzymes to which these cofactors were bound. The intensity ratio of NADH and FAD of cancerous cells was also shown to be larger than that of normal cells.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture