  • Search Results
  • The role of mitochondrial bioenergetics and oxidative stress in depressive behavior in recurrent concussion model in mice.
The role of mitochondrial bioenergetics and oxidative stress in depressive behavior in recurrent concussion model in mice.
Life sciences (2020-06-23)
Leandro Severo, Douglas Godinho, Felipe Machado, Diane Hartmann, Michele Rechia Fighera, Félix Alexandre Soares, Ana Flavia Furian, Mauro Schneider Oliveira, Luiz Fernando Royes
ABSTRACT
Traumatic brain injury (TBI) is a public health problem in which even though 80 to 90% of cases are considered mild, usually starts a sequence of neurological disorders that can last a considerable time. Most of the research of this injury has been focused on oxidative stress and functional deficits; however, mechanisms that underlie the development of neuropsychiatric disorders remain little researched. Due to this, the present authors decided to investigate whether recurrent concussion protocols alter depressive-like phenotype behavior, and whether mitochondria play an indispensable role in this behavior or not. The experimental data revealed, for the first time, that the present protocol of recurrent concussions (4, 7, and 10 injuries) in mice did not alter immobility time during tail suspension tests (TSTs), but decreased hippocampal mitochondrial respiration and increased expression of proteins such as nuclear factor erythroid 2-related factor 2 (Nrf2) and superoxide (SOD2). This experimental data suggests that bioenergetic changes elicited by recurrent concussion did not induce depressive-like behavior, but activated the transcription factor of responsive antioxidant elements (ARE) that delay or prevent secondary cascades in this neurological disease.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable for electrophoresis, 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
L-(−)-Malic acid, ≥95% (titration)
Sigma-Aldrich
(−)-Epinephrine (+)-bitartrate salt, solid
Supelco
Isovitexin, analytical standard