Merck
Fractionation of the Coxiella burnetii parasitophorous vacuole.
Methods in molecular biology (Clifton, N.J.) (2008-04-22)
Dale Howe, Robert A Heinzen
ABSTRACT
Coxiella burnetii is a bacterial obligate intracellular pathogen that replicates within a spacious parasitophorous vacuole (PV) with lysosomal characteristics. The pathogen actively participates in the biogenesis of its PV by synthesizing proteins that mediate vesicular interactions. Both C. burnetii and host factors that regulate PV formation are likely localized to the PV membrane, and their identification would be aided by an efficient method for isolating the C. burnetii vacuole. To this end, we developed a method to separate intact PV from host cell material that relies on fusion of the vacuole with latex bead-containing phagosomes (LBP). Sequestration of latex beads by the C. burnetii PV increases the vacuole's buoyant density and facilitates its fractionation on a sucrose step gradient. Transmission electron microscopy confirms the isolation of intact PV-containing latex beads from infected MH-S murine alveolar macrophage-like cells. Immunoblotting demonstrates that C. burnetii PV lysates are dramatically enriched for the late endosome/lysosome markers LAMP-1 and LAMP-2 when compared to total host cell lysates. Conversely, PV preparations are devoid of p62 and GM130, markers of the nucleus and Golgi apparatus, respectively, indicating effective separation of the vacuole from these host cell compartments. Two-dimensional gel electrophoresis and immunoblotting reveal distinct protein differences between C. burnetii PV and LBP. Identification of proteins unique to the PV membrane will yield important insight into C. burnetii-host interactions.
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
TWEEN® 20, viscosity 250-450 mPa.s (25 °C)
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid