  • Search Results
  • The effects of exogenous xylanase supplementation on the in vivo generation of xylooligosaccharides and monosaccharides in broilers fed a wheat-based diet.
The effects of exogenous xylanase supplementation on the in vivo generation of xylooligosaccharides and monosaccharides in broilers fed a wheat-based diet.
British poultry science (2020-07-21)
T Dale, I Hannay, M R Bedford, G A Tucker, J M Brameld, T Parr
ABSTRACT
1. This study quantified xylanase-induced changes in soluble monosaccharides, xylooligosaccharides (XOS) and volatile fatty acid (VFA) contents of the different sections of the gastrointestinal tract (GIT) and whether these were related to altered bird performance. 2. An in vitro digestion of the wheat-based diet was carried out with the xylanase (Econase XT at 16,000BXU/kg diet) to compare the in vitro and in vivo generation of these XOS and monosaccharides. For the in vivo study, 80 male Ross 508 b roiler chicks were split into two groups fed a wheat-based diet with or without Econase XT (16,000BXU/kg diet) for 21 days. 3. There were no effects of Econase XT inclusion on growth performance characteristics, likely a result of the high-quality wheat diet, the corresponding high performance of the control group (FCR average of 1.45 in controls) and the relatively young age of the birds (from four to 26 days of age). 4. Econase XT supplementation increased the xylotetraose (X4) content in the colon (P = 0.046, enzyme x GIT section interaction) and the xylose contents in the colon and caeca (P < 0.001, enzyme x GIT section interaction). 5. The trend for increased acetate production in the caeca of Econase XT treated birds (P = 0.062) suggested that the XOS generated were subsequently fermented in the caeca, potentially impacting upon the types of microbiota present. 6. The present study suggested that wheat arabinoxylan degradation was enhanced by xylanase supplementation, which may have increased the production of beneficial volatile fatty acids (VFA) in the caeca, and thereby potentially modulated the caecal microbiome, but without affecting bird performance at this early age.
Sigma-Aldrich
Xylobiose, ≥90% (HPLC)