  • Search Results
  • Synthesis of all-trans arachidonic acid and its effect on rabbit platelet aggregation.
Synthesis of all-trans arachidonic acid and its effect on rabbit platelet aggregation.
Bioorganic & medicinal chemistry letters (2005-05-07)
Dimitris Anagnostopoulos, Chryssostomos Chatgilialoglu, Carla Ferreri, Abdelouahid Samadi, Athanassia Siafaka-Kapadai
ABSTRACT
A simple and high-yielding method to convert natural all-cis PUFA derivatives to the corresponding all-trans geometrical isomers is described. The method is based on the thiyl radical-catalyzed cis-trans isomerization. The all-trans isomer of arachidonic acid was found to cause rabbit platelet aggregation at concentrations higher than 0.1 mM and inhibition of PAF-induced platelet aggregation in a concentration dependent manner with an IC(50) in the micromolar range.
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)