  • Search Results
  • Multiple N-methylation by a designed approach enhances receptor selectivity.
Multiple N-methylation by a designed approach enhances receptor selectivity.
Journal of medicinal chemistry (2007-11-02)
Jayanta Chatterjee, Oded Ovadia, Grit Zahn, Luciana Marinelli, Amnon Hoffman, Chaim Gilon, Horst Kessler
ABSTRACT
An unselective cyclic peptide integrin ligand was sequentially N-methylated by a designed approach, where only the externally oriented (solvent exposed) amide bonds were N-methylated. The N-methylation resulted in tremendous enhancement in selectivity among the different integrin receptor subtypes (alpha5beta1, alphavbeta3, and alphaIIbbeta3). Conformational and docking studies were performed, which suggested that the receptor selectivity is principally caused by reduced backbone flexibility due to N-methylation.
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Amoxicillin, potency: ≥900 μg per mg
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Supelco
Antipyrine, analytical standard
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination