  • Search Results
  • Synthesis and antimicrobial activity of some new thiazolyl thiazolidine-2,4-dione derivatives.
Synthesis and antimicrobial activity of some new thiazolyl thiazolidine-2,4-dione derivatives.
Bioorganic & medicinal chemistry (2007-07-10)
Oya Bozdağ-Dündar, Ozen Ozgen, Arzu Menteşe, Nurten Altanlar, Onur Atli, Engin Kendi, Rahmiye Ertan
ABSTRACT
In this study, a series of thiazolyl thiazolidine-2,4-dione derivatives (Va-f and VIa-f) were synthesized and evaluated for their antibacterial and antifungal activities against Staphylococcus aureus (ATCC 25923), methicillin resistant S. aureus (MRSA ATCC 43300), methicillin resistant S. aureus (MRSA isolate), and Escherichia coli (ATCC 23556) and C. albicans (ATCC10145). All the compounds were found active against used microorganisms.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard