  • Search Results
  • miR-183/96/182 cluster is an important morphogenetic factor targeting PAX6 expression in differentiating human retinal organoids.
miR-183/96/182 cluster is an important morphogenetic factor targeting PAX6 expression in differentiating human retinal organoids.
Stem cells (Dayton, Ohio) (2020-09-03)
Lucie Peskova, Denisa Jurcikova, Tereza Vanova, Jan Krivanek, Michaela Capandova, Zuzana Sramkova, Jana Sebestikova, Magdalena Kolouskova, Hana Kotasova, Libor Streit, Tomas Barta
ABSTRACT
MicroRNAs (miRNAs), a class of small, noncoding RNA molecules represent important regulators of gene expression. Recent reports have implicated their role in the cell specification process acting as "fine-tuners" to ensure the precise gene expression at the specific stage of cell differentiation. Here, we used retinal organoids differentiated from human pluripotent stem cells (hPSCs) as a model to closely investigate the role of a sensory organ-specific and evolutionary conserved miR-183/96/182 cluster. Using a miRNA tough decoy approach, we inhibited the miR-183/96/182 cluster in hPSCs. Inhibition of the miRNA cluster resulted in an increased expansion of neuroepithelium leading to abnormal "bulged" neural retina in organoids, associated with upregulation of neural-specific and retinal-specific genes. Importantly, we identified PAX6, a well-known essential gene in neuroectoderm specification, as a target of the miR-183/96/182 cluster members. Taken together, the miR-183/96/182 cluster not only represents an important regulator of PAX6 expression, but it also plays a crucial role in retinal tissue morphogenesis.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, for molecular biology
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, FITC conjugate, clone 4G10®, Upstate®, from mouse