  • Search Results
  • Evidence for moonlighting functions of the θ subunit of Escherichia coli DNA polymerase III.
Evidence for moonlighting functions of the θ subunit of Escherichia coli DNA polymerase III.
Journal of bacteriology (2014-01-01)
M Dietrich, L Pedró, J García, M Pons, M Hüttener, S Paytubi, C Madrid, A Juárez
ABSTRACT
The holE gene is an enterobacterial ORFan gene (open reading frame [ORF] with no detectable homology to other ORFs in a database). It encodes the θ subunit of the DNA polymerase III core complex. The precise function of the θ subunit within this complex is not well established, and loss of holE does not result in a noticeable phenotype. Paralogs of holE are also present on many conjugative plasmids and on phage P1 (hot gene). In this study, we provide evidence indicating that θ (HolE) exhibits structural and functional similarities to a family of nucleoid-associated regulatory proteins, the Hha/YdgT-like proteins that are also encoded by enterobacterial ORFan genes. Microarray studies comparing the transcriptional profiles of Escherichia coli holE, hha, and ydgT mutants revealed highly similar expression patterns for strains harboring holE and ydgT alleles. Among the genes differentially regulated in both mutants were genes of the tryptophanase (tna) operon. The tna operon consists of a transcribed leader region, tnaL, and two structural genes, tnaA and tnaB. Further experiments with transcriptional lacZ fusions (tnaL::lacZ and tnaA::lacZ) indicate that HolE and YdgT downregulate expression of the tna operon by possibly increasing the level of Rho-dependent transcription termination at the tna operon's leader region. Thus, for the first time, a regulatory function can be attributed to HolE, in addition to its role as structural component of the DNA polymerase III complex.
Sigma-Aldrich
Complete Whole Transcriptome Amplification Kit, DNA polymerase included, Complete Kit with optimized enzyme to amplify total RNA in <4 hours, no 3′ bias