  • Search Results
  • Structure-brain exposure relationships in rat and human using a novel data set of unbound drug concentrations in brain interstitial and cerebrospinal fluids.
Structure-brain exposure relationships in rat and human using a novel data set of unbound drug concentrations in brain interstitial and cerebrospinal fluids.
Journal of medicinal chemistry (2009-09-22)
Markus Fridén, Susanne Winiwarter, Gunilla Jerndal, Ola Bengtsson, Hong Wan, Ulf Bredberg, Margareta Hammarlund-Udenaes, Madeleine Antonsson
ABSTRACT
New experimental methodologies were applied to measure the unbound brain-to-plasma concentration ratio (K(p,uu,brain)) and the unbound CSF-to-plasma concentration ratio (K(p,uu,CSF)) in rats for 43 structurally diverse drugs. The relationship between chemical structure and K(p,uu,brain) was dominated by hydrogen bonding. Contrary to popular understanding based on the total brain-to-plasma concentration ratio (logBB), lipophilicity was not a determinant of unbound brain exposure. Although changing the number of hydrogen bond acceptors is a useful design strategy for optimizing K(p,uu,brain), future improvement of in silico prediction models is dependent on the accommodation of active drug transport. The structure-brain exposure relationships found in the rat also hold for humans, since the rank order of the drugs was similar for human and rat K(p,uu,CSF). This cross-species comparison was supported by K(p,uu,CSF) being within 3-fold of K(p,uu,brain) in the rat for 33 of 39 drugs. It was, however, also observed that K(p,uu,CSF) overpredicts K(p,uu,brain) for highly effluxed drugs, indicating lower efflux capacity of the blood-cerebrospinal fluid barrier compared to the blood-brain barrier.
Product Description
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Sigma-Aldrich
Rifampicin, ≥97% (HPLC), powder
Sigma-Aldrich
Gabapentin, solid
Sigma-Aldrich
(±)-Thiopental
Sigma-Aldrich
(±)-Baclofen, ≥98% (TLC), solid
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Supelco
Norfloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥97% (HPLC), crystalline
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Salicylic acid, certified reference material, TraceCERT®