  • Search Results
  • Carbonic anhydrase activators: activation of the beta-carbonic anhydrase Nce103 from the yeast Saccharomyces cerevisiae with amines and amino acids.
Carbonic anhydrase activators: activation of the beta-carbonic anhydrase Nce103 from the yeast Saccharomyces cerevisiae with amines and amino acids.
Bioorganic & medicinal chemistry letters (2009-02-24)
Semra Isik, Feray Kockar, Meltem Aydin, Oktay Arslan, Ozen Ozensoy Guler, Alessio Innocenti, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
The protein encoded by the Nce103 gene of Saccharomyces cerevisiae, a beta-carbonic anhydrase (CA, EC 4.2.1.1) designated as scCA, was investigated for its activation with amines and amino acids. scCA was poorly activated by amino acids such as l-/d-His, Phe, DOPA, Trp (K(A)s of 82-90 microM) and more effectively activated by amines such as histamine, dopamine, serotonin, pyridyl-alkylamines, aminoethyl-piperazine/morpholine (K(A)s of 10.2-21.3 microM). The best activator was l-adrenaline, with an activation constant of 0.95 microM. This study may help to better understand the catalytic/activation mechanisms of the beta-CAs and eventually to design modulators of CA activity for similar enzymes present in pathogenic fungi, such as Candida albicans and Cryptococcus neoformans.
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
Supelco
L-Tryptophan, certified reference material, TraceCERT®
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
SAFC
L-Tyrosine
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Sigma-Aldrich
L-Phenylalanine, 99%
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
Supelco
L-Phenylalanine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Supelco
L-Phenylalanine, analytical standard, for Nitrogen Determination According to Kjeldahl Method