  • Search Results
  • Synthesis of 6-aminomethyl derivatives of benzopyran-4-one with dual biological properties: anti-inflammatory-analgesic and antimicrobial.
Synthesis of 6-aminomethyl derivatives of benzopyran-4-one with dual biological properties: anti-inflammatory-analgesic and antimicrobial.
European journal of medicinal chemistry (2009-08-29)
S M Hasan, M M Alam, Asif Husain, Suruchi Khanna, Mymoona Akhtar, M S Zaman
ABSTRACT
A series of 6-aminomethyl-2-aryl-1-benzopyran-4-one derivatives (10-24) were synthesized. The compounds were tested for anti-inflammatory, analgesic, ulcerogenic and lipid peroxidation actions. Among the tested compounds, six compounds 11, 13, 16, 18, 21 and 23 showed higher degree of anti-inflammatory activity (>75% activity of standard drug ibuprofen). In addition to remarkable anti-inflammatory activity, analgesic activity was found to be comparable with that of the standard drug ibuprofen. Compounds 16 and 21 showed a significant GI protection (with respect to ulcerogenesis) and a marked decrease in lipid peroxidation values whereas compounds 11 and 16 were found to possess antimicrobial activity against Staphylococcus aureus, Escherichia coli, Rhizopus oryza and Penicillum citrum with an MIC of 10 microg/mL.
Product Description
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Norfloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Supelco
Ibuprofen