  • Search Results
  • Messenger RNA 5' NAD+ Capping Is a Dynamic Regulatory Epitranscriptome Mark That Is Required for Proper Response to Abscisic Acid in Arabidopsis.
Messenger RNA 5' NAD+ Capping Is a Dynamic Regulatory Epitranscriptome Mark That Is Required for Proper Response to Abscisic Acid in Arabidopsis.
Developmental cell (2020-12-09)
Xiang Yu, Matthew R Willmann, Lee E Vandivier, Sophie Trefely, Marianne C Kramer, Jeffrey Shapiro, Rong Guo, Eric Lyons, Nathaniel W Snyder, Brian D Gregory
ABSTRACT
Although eukaryotic messenger RNAs (mRNAs) normally possess a 5' end N7-methyl guanosine (m7G) cap, a non-canonical 5' nicotinamide adenine dinucleotide (NAD+) cap can tag certain transcripts for degradation mediated by the NAD+ decapping enzyme DXO1. Despite this importance, whether NAD+ capping dynamically responds to specific stimuli to regulate eukaryotic transcriptomes remains unknown. Here, we reveal a link between NAD+ capping and tissue- and hormone response-specific mRNA stability. In the absence of DXO1 function, transcripts displaying a high proportion of NAD+ capping are instead processed into RNA-dependent RNA polymerase 6-dependent small RNAs, resulting in their continued turnover likely to free the NAD+ molecules. Additionally, the NAD+-capped transcriptome is significantly remodeled in response to the essential plant hormone abscisic acid in a mechanism that is primarily independent of DXO1. Overall, our findings reveal a previously uncharacterized and essential role of NAD+ capping in dynamically regulating transcript stability during specific physiological responses.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Phenol – chloroform – isoamyl alcohol mixture, BioUltra, for molecular biology, 25:24:1
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Chloroform:Isoamyl alcohol 24:1, suitable for nucleic acid purification
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration)
Sigma-Aldrich
Tris(3-hydroxypropyltriazolylmethyl)amine, 95%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
4-Pentyn-1-ol, 97%
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)
Sigma-Aldrich
Albumin, Acetylated from bovine serum, protease free, for molecular biology
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, ≥98%