  • Search Results
  • Synthesis and biological activities of novel amine-derived bis-azoles as potential antibacterial and antifungal agents.
Synthesis and biological activities of novel amine-derived bis-azoles as potential antibacterial and antifungal agents.
European journal of medicinal chemistry (2010-07-06)
Bo Fang, Cheng-He Zhou, Xian-Cai Rao
ABSTRACT
A series of novel amine-derived bis-azole compounds were designed by the systematical structural modification of Fluconazole and synthesized by a convenient and efficient method, and the antimicrobial activities for all prepared compounds were evaluated in vitro against six representative bacterial strains and two fungal strains. Bioactive results indicated that some synthesized compounds exhibited moderate or even better activities in comparison with the reference drugs. Especially, bis-imidazole 5b and its salts gave significant antibacterial efficacy against all tested bacteria strains including MRSA, while bis-triazoles 4b-c and their corresponding salts exhibited better activities against Candida albicans, Bacillus proteus than standard drugs Fluconazole and Norfloxacin respectively. Unexpectedly, bis-bromides 3a-f presented excellent activities against all tested microbial strains.
Product Description
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Norfloxacin, VETRANAL®, analytical standard