  • Search Results
  • Biomimetic microenvironmental preconditioning enhance neuroprotective properties of human mesenchymal stem cells derived from Wharton's Jelly (WJ-MSCs).
Biomimetic microenvironmental preconditioning enhance neuroprotective properties of human mesenchymal stem cells derived from Wharton's Jelly (WJ-MSCs).
Scientific reports (2020-10-11)
Wioletta Lech, Anna Sarnowska, Zuzanna Kuczynska, Filip Dabrowski, Anna Figiel-Dabrowska, Krystyna Domanska-Janik, Leonora Buzanska, Marzena Zychowicz
ABSTRACT
Tuning stem cells microenvironment in vitro may influence their regenerative properties. In this study Wharton's Jelly-derived mesenchymal stem cells (WJ-MSCs) were encapsulated in 3D hydrogels derived from human fibrin (FB) or platelet lysate (PL) and the oxygen level was adjusted to physiological normoxia (5% O2). The influence of the type of the scaffold and physiological normoxia conditions was tested on the WJ-MSCs' survivability, proliferation, migratory potential, the level of expression of selected trophic factors, cytokines, and neural markers. Encapsulated WJ-MSCs revealed high survivability, stable proliferation rate, and ability to migrate out of the hydrogel and the up-regulated expression of all tested factors, as well as the increased expression of neural differentiation markers. Physiological normoxia stimulated proliferation of encapsulated WJ-MSCs and significantly enhanced their neuronal, but not glial, differentiation. Ex vivo studies with indirect co-culture of organotypic hippocampal slices and cell-hydrogel bio-constructs revealed strong neuroprotective effect of WJ-MSCs against neuronal death in the CA1 region of the rat hippocampus. This effect was potentiated further by FB scaffolds under 5% O2 conditions. Our results indicating significant effect of oxygen and 3D cytoarchitecture suggest the urgent need for further optimization of the microenvironmental conditions to improve therapeutical competence of the WJ-MSCs population.
Product Description
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile