  • Search Results
  • Optimized Nile Red efflux assay of AcrAB-TolC multidrug efflux system shows competition between substrates.
Optimized Nile Red efflux assay of AcrAB-TolC multidrug efflux system shows competition between substrates.
Antimicrobial agents and chemotherapy (2010-07-08)
Jürgen A Bohnert, Brian Karamian, Hiroshi Nikaido
ABSTRACT
AcrAB-TolC is the major constitutively expressed efflux pump system that provides resistance to a variety of antimicrobial agents and dyes in Escherichia coli. However, no systematically optimized real-time dye efflux assay has been published for the measurement of its activity and for detection of possible competition between substrates. Here, we report on the development of such an assay using a lipophilic dye, Nile Red. Energy-depleted cells were loaded with the dye in the presence of low (10 microM or less) concentrations of the proton conductor carbonyl cyanide m-chlorophenylhydrazone (CCCP). The CCCP was then removed, and efflux was triggered by energization with glucose. Various known efflux pump inhibitors and antimicrobials were checked for the ability to slow down Nile Red efflux, presumably through competition. Besides the known inhibitors Phe-Arg-beta-naphthylamide and 1-naphthyl-methylpiperazine, several tetracyclic compounds (doxorubicin, minocycline, chlortetracycline, doxycycline, and tetracycline) and tetraphenylphosphonium chloride were found to interfere with dye efflux. This inhibition could not be explained by the depletion of proton motive force. None of the other tested antimicrobials, including macrolides, fluoroquinolones, and beta-lactams, had any impact on Nile Red efflux, even at concentrations of up to 1 mM.
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Supelco
Tetraphenylphosphonium chloride, for the spectrophotometric det. of Bi, Co, ≥97.0%
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Tetraphenylphosphonium chloride, 98%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Deoxycholic acid, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Sigma-Aldrich
Rifampicin, ≥97% (HPLC), powder
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Nalidixic acid, ≥98%
Sigma-Aldrich
Tetraphenylphosphonium bromide, 97%
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Norfloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥97% (HPLC), crystalline