  • Search Results
  • Tryptophan Attenuates the Effects of OTA on Intestinal Morphology and Local IgA/IgY Production in Broiler Chicks.
Tryptophan Attenuates the Effects of OTA on Intestinal Morphology and Local IgA/IgY Production in Broiler Chicks.
Toxins (2020-12-31)
Fernando Galdino Ricci, Leticia Rodrigues Terkelli, Emerson José Venancio, Larissa Justino, Beatriz Queiroz Dos Santos, Ana Angelita Sampaio Baptista, Alexandre Oba, Bianca Dorana de Oliveira Souza, Ana Paula Frederico Rodrigues Loureiro Bracarense, Elisa Yoko Hirooka, Eiko Nakagawa Itano
ABSTRACT
Ochratoxin A (OTA) is a mycotoxin produced by species of Penicillium and Aspergillus that can contaminate products of plant origin that are used as animal feed. Through oral exposure, this mycotoxin primarily affects the chicken gastrointestinal system. The present study evaluated the intestinal toxic effects of OTA and the introduction of L-tryptophan to alleviate these effects in chickens. One-day-old chicks were exposed to a single OTA dose (1.4 mg/kg body weight-b.w.) and treated with or without four daily doses of L-tryptophan (100 mg/kg b.w.). Duodenal villus height/crypt depth, fecal immunoglobulin A/immunoglobulin Y (IgA/IgY) levels, and duodenal positive immunoglobulin A cells (IgA+) were evaluated by histology, ELISA, and immunohistochemistry, respectively, on the 14th day. There were significant changes in the duodenal villus height, crypt depth, and levels of fecal IgA/IgY and duodenal IgA+ cells (p < 0.05) in groups exposed to OTA. On the other hand, groups exposed to OTA and treated with L-tryptophan showed similar levels of villus height, IgA/IgY levels, and duodenal IgA+ cells to those of the control group (p > 0.05). In conclusion, exposure to a single dose of OTA orally induces changes in intestinal morphology, levels of IgA/IgY antibodies, and IgA+ cells. Thus, treatment with L-tryptophan may be a valid alternative means to reduce the harmful effects of OTA on the intestinal mucosa, which requires further study.
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Anti-Chicken IgY (IgG) (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution