  • Search Results
  • Prevalence and clonal characterization of Streptococcus pyogenes clinical isolates with reduced fluoroquinolone susceptibility in Spain.
Prevalence and clonal characterization of Streptococcus pyogenes clinical isolates with reduced fluoroquinolone susceptibility in Spain.
Antimicrobial agents and chemotherapy (2009-10-07)
Milagrosa Montes, Esther Tamayo, Beatriz Orden, Julián Larruskain, Emilio Perez-Trallero
ABSTRACT
The aim of this study was to determine the prevalence and characteristics of non-fluoroquinolone (FQ)-susceptible Streptococcus pyogenes isolates and to study their mechanisms of resistance. We performed a prospective prevalence study with 468 isolates collected from 2005 to 2007 and a retrospective study that was based on the examination of existing data collected from 1999 to 2008. The retrospective study included data for isolates with high-level resistance (HR) to ciprofloxacin (MIC >or= 32 microg/ml) (HR isolates) and isolates with the same emm types as those reported in the literature with low-level resistance (LR) to ciprofloxacin (MICs, 2 to 8 microg/ml) (LR isolates, n = 205). Genetic characterization of the isolates was performed by means of emm typing and multilocus sequence typing. The prevalence of LR ranged from 1.9% in 2005 to 30.8% in 2007. This increase was mainly due to the circulation of an emm6 subtype (emm6.4) that represented 77.1% of the LR isolates in 2007. Notably, another emm6 subtype, also detected in 2007 (emm6.37), showed coresistance to 14- and 15-membered macrolides mediated by the mefA gene. Only three HR isolates were detected (isolates emm68.1/ST247/T3,13,B3264, emm77/ST399/T28, and emm28/ST52/T28), and all were identified in the retrospective study. Overall, the 673 isolates represented 25 emm types. All LR isolates were clustered into two emm types: emm6 (six emm6 subtypes) and emm75. All the 156 emm6 isolates had LR, harbored the Ser79/Ala mutation in the parC gene product, and had the same sequence type (ST), ST382. Most (21/33) of the emm75 isolates had LR, showed the Ser79/Phe plus Asp91/Asn double mutation in the parC gene product, and were ST150. The Asp91/Asn mutation by itself did not confer resistance to FQs.
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Norfloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)