  • Search Results
  • Green tea infusion reduces mercury bioaccessibility and dietary exposure from raw and cooked fish.
Green tea infusion reduces mercury bioaccessibility and dietary exposure from raw and cooked fish.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-09-06)
Patrícia Anacleto, Vera Barbosa, Ricardo N Alves, Ana Luísa Maulvault, Maria Rosário Bronze, António Marques
ABSTRACT
Human exposure to mercury (Hg) and methylmercury (MeHg) through the ingestion of seafood raises human health-related concerns. In contrast, green tea has health benefits and its consumption potentially reduces bioaccessibility of dietary Hg. The present study aimed to assess the effect of green tea in total mercury (THg) and MeHg bioaccessibility in raw and cooked marine fish species commonly having high Hg levels. Preliminary results demonstrated that significantly higher reductions of bioaccessible THg were attained after the co-ingestion of green tea infusion (1 cup or more) in the oral and intestinal phases. Overall, the present findings clearly show that the co-ingestion of green tea along with seafood grilling strongly reduces THg and MeHg bioaccessibility in all fish species and consequently diminishes the probability of exceeding MeHg provisional tolerable weekly intakes through the consumption of these species with high Hg levels. Such results point out the need to better understand the beneficial/preventive role of green tea infusions and other food processing techniques in bioaccessibility reduction of other chemical contaminants present in food products. Such information is certainly useful to help consumers to wisely select their food, and to enable food safety authorities to integrate such information in risk assessment.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Mucin from porcine stomach, Type II
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, 100-650 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
(+)-Catechin hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
Potassium thiocyanate, BioXtra, ≥99.0%
Sigma-Aldrich
Pancreatin from porcine pancreas, ≥3 × USP specifications
Sigma-Aldrich
D-(+)-Glucosamine hydrochloride, ≥99%, BioReagent, suitable for cell culture
Sigma-Aldrich
BIO, ≥98% (HPLC)