In the study of previously reported modulators of CFTR chloride channels that are cyclic methylglyoxal (MG) diadducts (CMGD) to aromatic α-aminoazaheterocycles, we optimized a new expeditious one pot route for preparing in water novel aromatic polycyclic azaheterocycles and described 5-pyrimidinols antioxidants through the formation of 2-oxoaldehyde diadducts to aromatic α-aminoazaheterocycles, amidines, guanidines and thiourea. In regard to the importance as biomarkers of diabetic complications of the 5-pyrimidinols "argpyrimidines" formed in proteins from MG and arginine residues, we demonstrated that argpyrimidines are slowly formed under physiological conditions from CMGD to arginine derivatives according to the synthesis route described. Among the 5-pyrimidinol derivatives prepared, two polycyclic derivatives appeared to inhibit strongly the activity of CFTR channels in wt-CHO cells.