  • Search Results
  • Synthesis and antibacterial activity of novel 4″-O-benzimidazolyl clarithromycin derivatives.
Synthesis and antibacterial activity of novel 4″-O-benzimidazolyl clarithromycin derivatives.
European journal of medicinal chemistry (2011-04-29)
Chao Cong, Haiyang Wang, Yue Hu, Chen Liu, Siti Ma, Xin Li, Jichao Cao, Shutao Ma
ABSTRACT
Novel 4″-O-benzimidazolyl clarithromycin derivatives were designed, synthesized and evaluated for their in vitro antibacterial activities. These benzimidazolyl derivatives exhibited excellent activity against erythromycin-susceptible strains better than the references, and some of them showed greatly improved activity against erythromycin-resistant strains. Compounds 16 and 17, which have the terminal 2-(4-methylphenyl)benzimidazolyl and 2-(2-methoxyphenyl)benzimidazolyl groups on the C-4″ bishydrazide side chains, were the most active against erythromycin-resistant Staphylococcus pneumoniae expressing the erm gene and the mef gene. In addition, compound 17 exhibited the highest activity against erythromycin-susceptible S. pneumoniae ATCC49619 and Staphylococcus aureus ATCC25923 as well. It is worth noting that the 4″-O-(2-aryl)benzimidazolyl derivatives show higher activity against erythromycin-susceptible and erythromycin-resistant strains than the 4″-O-(2-alkyl)benzimidazolyl derivatives.
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.